Learn More
Cayman Chemical CItrulInated GFAPhumnre 100ug
SDP
Catalog No. 502937063
Encompass_Preferred
Source: Recombinant N-terminal histidine-tagged GFAP expressed in E. coli. Citrullinated with recombinant human PAD2 • Amino acids: 2-432 • MW: 51.9 kDa
Catalog No. 50-293-7063 Supplier Cayman Chemical Supplier No. 28622100 UG
Source: Recombinant N-terminal histidine-tagged GFAP expressed in E. coli. Citrullinated with recombinant human PAD2 • Amino acids: 2-432 • MW: 51.9 kDa