Learn More
Cayman Chemical Citrullinated GRP78 (R368) Polyclonal Antibody, 1 ea
SDP
Catalog No. 502934240
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to an internal region of human GRP78 with a citrulline at residue 368 • Host: Rabbit • Cross Reactivity: (+) Citrullinated GRP78 and citrullinated HSP70; (-) Unmodified GRP78, unmodified HSP70, and all other citrullinated proteins • Species Reactivity: (+) Human citrullinated GRP78 • Applications: ELISA and WB
Catalog No. 50-293-4240 Supplier Cayman Chemical Supplier No. 242871 EA
Immunogen: Synthetic peptide corresponding to an internal region of human GRP78 with a citrulline at residue 368 • Host: Rabbit • Cross Reactivity: (+) Citrullinated GRP78 and citrullinated HSP70; (-) Unmodified GRP78, unmodified HSP70, and all other citrullinated proteins • Species Reactivity: (+) Human citrullinated GRP78 • Applications: ELISA and WB