Learn More
Cayman Chemical Citrullinated Hsp70 (R155) Polyclonal Antibody, 1 ea
SDP
Catalog No. 502933922
Encompass_Preferred
Immunogen: Peptide from the internal region of human Hsp70 containing a citrulline at residue 155 • Host: Rabbit • Cross Reactivity: (+) Citrullinated Hsp70, Citrullinated GRP78; (-) Hsp70, other citrullinated Hsp proteins • Species Reactivity: (+) Human; other species not tested • Applications: ELISA and WB
Catalog No. 50-293-3922 Supplier Cayman Chemical Supplier No. 244971 EA
Immunogen: Peptide from the internal region of human Hsp70 containing a citrulline at residue 155 • Host: Rabbit • Cross Reactivity: (+) Citrullinated Hsp70, Citrullinated GRP78; (-) Hsp70, other citrullinated Hsp proteins • Species Reactivity: (+) Human; other species not tested • Applications: ELISA and WB