Learn More
Cayman Chemical CItrulInated InTRleukIn6 100ug
SDP
Catalog No. 502989142
Encompass_Preferred
Source: Recombinant human N-terminal His-tagged IL-6 expressed in HEK293 cells, citrullinated by PAD4 • Amino acids: 30-212 • MW: 20.9 kDa
Catalog No. 50-298-9142 Supplier Cayman Chemical Supplier No. 30173100 UG
Source: Recombinant human N-terminal His-tagged IL-6 expressed in HEK293 cells, citrullinated by PAD4 • Amino acids: 30-212 • MW: 20.9 kDa