Learn More
Cayman Chemical CItrulInated InTRleukIn6 P 1mL
SDP
Catalog No. 502911607
Encompass_Preferred
Immunogen: Synthetic citrullinated peptide corresponding to an internal region of human IL-6 • Host: Rabbit • Species Reactivity: (+) Human; (-) Others not tested • Cross Reactivity: (+) Citrullinated IL-6 • Applications: ELISA, WB • MW = ~24 kDa
Catalog No. 50-291-1607 Supplier Cayman Chemical Supplier No. 361781 ML
Immunogen: Synthetic citrullinated peptide corresponding to an internal region of human IL-6 • Host: Rabbit • Species Reactivity: (+) Human; (-) Others not tested • Cross Reactivity: (+) Citrullinated IL-6 • Applications: ELISA, WB • MW = ~24 kDa