Learn More
Cayman Chemical CItrulInated l37 MoncLNa 300ug
SDP
Catalog No. 502934951
Encompass_Preferred
Immunogen: Citrullinated human LL-37 peptide • Host: Mouse• Species Reactivity: (+) Human • Cross Reactivity: (+) Citrullinated LL-37; (-) Native LL-37 • Applications: ELISA, WB • MW = ~9 kDa
Catalog No. 50-293-4951 Supplier Cayman Chemical Supplier No. 26739300 UG
Immunogen: Citrullinated human LL-37 peptide • Host: Mouse• Species Reactivity: (+) Human • Cross Reactivity: (+) Citrullinated LL-37; (-) Native LL-37 • Applications: ELISA, WB • MW = ~9 kDa