Learn More
Cayman Chemical Citrullinated Vimentin Monoclonal Antibody (Clone 12G11), 100 µg
SDP
Supplier:  Cayman Chemical 22054100
Encompass_Preferred
Immunogen: Citrullinated peptide sequence from an internal region of human vimentin • Clone Designation: 12G11 • Host: Mouse • Cross Reactivity: (+) Citrullinated vimentin; all others not tested though sequence conservation of >95% with bovine, pig, mouse, and rat indicates potential species reactivity, (-) Unmodified protein • Species Reactivity: (+) Human citrullinated vimentin • Isotype: IgG1 • Applications: ELISA and WB,
Catalog No. NC1370901