Learn More
Cayman Chemical Human Fibrinogen (PAD4 Citrullinated), 1 mg
SDP
Supplier:  Cayman Chemical 4000761
Encompass_Preferred
Source: Fibrinogen, purified from human plasma, that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV. Citrullinated with human recombinant PAD4 Uncitrullinated Fibrinogen Mr: chain isoform 1 (95 kDa), chain isoform 2 (69.8 kDa), chain (55.9kDa) and isoform -B chain (51.5 kDa).
Catalog No. NC0771277
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.