Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Citrulline (L) (Dietary Supplement Grade)
SDP

Catalog No. 14186238 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-238 1 kg
14-185-963 500 g
14-186-159 100 g
14-185-470 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-186-238 Supplier Medisca Supplier No. 118609
Get 1 Extra Chemical When You Buy 4
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Citrulline (L) (Dietary Supplement Grade) is white and comes as crystals or as a crystalline powder. It is practically odorless.

Citrulline (L) is white and comes as crystals or as a crystalline powder. It is practically odorless.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Alternative name(s): Citrulline

Specifications

Chemical Name or Material Citrulline (L) (Dietary Supplement Grade)
Packaging Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 372-75-8
Type Drug Standard
Molecular Weight (g/mol) 175.187
Quantity 1 kg
Percent Purity 99.5 to 101.0 %
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.