Learn More
Medchemexpress LLC Citrulline-specific probe-rhodamine (hydrate) | 2309313-01-5 | 731.75 | 250 UG
SDP
Catalog No. 5000429320
Encompass_Preferred
Citrulline-specific probe-rhodamine hydrate is a specific probe for citrulline combined with a rhodamine fluorescent group. Citrulline is the hydrolysis product of arginine catalyzed by protein arginine deiminase (PAD). PAD is abnormally activated in many diseases, leading to increased citrulline levels. This probe is a biological tool that can identify diseases showing abnormal increases in PAD activity and may be effectively used in animal models of ulcerative colitis.
  • Specific probe for citrulline
  • Combined with a rhodamine fluorescent group
  • Identifies diseases with abnormal increases in PAD activity
  • May be effectively used in animal models of ulcerative colitis
  • For research use only
Catalog No. 50-004-29320 Supplier Medchemexpress LLC Supplier No. 250UGHYD1377
Citrulline-specific probe-rhodamine hydrate is a specific probe for citrulline combined with a rhodamine fluorescent group. Citrulline is the hydrolysis product of arginine catalyzed by protein arginine deiminase (PAD). PAD is abnormally activated in many diseases, leading to increased citrulline levels. This probe is a biological tool that can identify diseases showing abnormal increases in PAD activity and may be effectively used in animal models of ulcerative colitis.
  • Specific probe for citrulline
  • Combined with a rhodamine fluorescent group
  • Identifies diseases with abnormal increases in PAD activity
  • May be effectively used in animal models of ulcerative colitis
  • For research use only