Learn More
Cayman Chemical CItrulntd Myeln BasIc Pr 500uL
SDP
Catalog No. 502934952
Encompass_Preferred
An orally bioavailable broad-spectrum triazole antifungal agent completely inhibits growth of T. rubrum M. audouini M. canis and T. verrucosum as well as some C. albicans and A. fumigatus strains and other fungi (100 g/ml) bacteriostatic against E. coli P. aeruginosa S. aureus and S. pyogenes (100 g/ml) eliminates vaginal C. albicans candidiasis infection in 97 of rats (1 topical) eliminates vaginal C. albicans candidiasis infection in 50 of rats (10 mg/kg oral) decreases synthesis of 14a-desmethyl sterols and increases synthesis of methylated sterols in C. albicans (IC50 3 nM)
Catalog No. 50-293-4952 Supplier Cayman Chemical Supplier No. 26742500 UL
An orally bioavailable broad-spectrum triazole antifungal agent completely inhibits growth of T. rubrum M. audouini M. canis and T. verrucosum as well as some C. albicans and A. fumigatus strains and other fungi (100 g/ml) bacteriostatic against E. coli P. aeruginosa S. aureus and S. pyogenes (100 g/ml) eliminates vaginal C. albicans candidiasis infection in 97 of rats (1 topical) eliminates vaginal C. albicans candidiasis infection in 50 of rats (10 mg/kg oral) decreases synthesis of 14a-desmethyl sterols and increases synthesis of methylated sterols in C. albicans (IC50 3 nM)