Learn More
Epredia™ Citrus Clearing Solvent
GSA_VA
Catalog No. 22050122 Shop All Epredia Products
Change view
Click to view available options
Quantity:
4 x 3.8L (4 x 1 gal.)
Catalog No. Quantity
22-050-122 4 x 3.8L (4 x 1 gal.)
1 options
This item is not returnable. View return policy
Catalog No. 22-050-122 Supplier Epredia™ Supplier No. 8301
This item is not returnable. View return policy
Replace xylene in both manual and automated system applications with Epredia™ Citrus Clearing Solvent.
  • D-limonene-based solvent has a slight citrus scent.
  • Higher flash point eliminates the need for storage in a flammable safety cabinet.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 3.8L (4 x 1 gal.)