Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ Citrus Ginger Foam Hand and Showerwash Refill
GREENER_CHOICESDP

Catalog No. p-7145678 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
1250 mL
For Use With (Equipment):
GOJO™ ADX-12™ Dispenser
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) For Use With (Equipment)
19-043-841 1250 mL GOJO™ ADX-12™ Dispenser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-043-841 Supplier GOJO™ Supplier No. 881303
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

All-in-one body wash, shampoo, and hand soap with a refreshing blend of citrus and ginger extracts.

  • Enriched with moisturizers, natural extracts and skin conditioners
  • Refill helps prevent contamination
  • Fresh dispensing valve with each refill
  • Maintains bottle shape longer while emptying
  • Removable pump for easier recycling
  • Fresh dispensing valve with each refill
TRUSTED_SUSTAINABILITY

Specifications

Product Type Body Wash
Color Yellow
Form Foam
Certifications/Compliance Meets UL EcoLogo Standard for Hand Cleaner UL 2784, Meets USDA Biobased Standard for Hand Cleaners
For Use With (Application) Wash
For Use With (Equipment) GOJO™ ADX-12™ Dispenser
Quantity 3/Pk.
Size 1.25 L
Volume (English) 42.27 oz.
Volume (Metric) 1250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.