Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citrus Red 2, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014996 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4996 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4996 Supplier MilliporeSigma Supelco Supplier No. 8977425MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Citrus red 2 is an artificial dye, which belongs to the mono-azo group of dyes. It may be used to color orange skins.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 6358-53-8
Physical Form Neat
Linear Formula C18H16N2O3
MDL Number MFCD01712266
Synonym 1-(2,5-Dimethoxyphenylazo)-2-naphthol
Type Analytical Standard
Quantity 25 mg
Formula Weight 308.33
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.