Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cramer Citrus Stool

Catalog No. 03155054
Change view
Click to view available options
Back Style:
Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Back Style
03-155-054 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-155-054 Supplier Cramer Supplier No. CTHU12
Add to Cart
Add to Cart

Fast-paced thinkers need seating that rolls from station to station, adjusts to every sit style, and wipes clean without the drama. Citrus solves it all – minimalist in style, maximum in utility.

Simple comfort The Flex Function back auto-adjusts leaving only one-touch height adjustment. Perch or sit Waterfall edge seat accommodates forward leaning postures. Easy to clean Seamless design with minimal joints keeps disinfection simple. The Citrus keeps things simple, balancing style with performance. Ah, what a refreshing take on lab and healthcare seating. Light and fresh A minimalist look with your choice of optional vibrant colors.

Features

  • Flex Function back and forward edge seat adapt and auto-adjust to individual user movements and support perching or sitting postures
  • One-touch height adjustment-is the only user-made adjustment necessary
  • Minimal joint hygienic design for easy cleaning and disinfection
  • Meets BIFMA Healthcare Furniture Cleanability Guidelines HCF 8.1-2019
  • ISO-Class 3 cleanroom rated
  • 300 lb. weight capacity
  • SCS Indoor Advantage Gold Certification and may help contribute toward LEED certification

Construction

  • Simple design and minimalist construction with few moving parts
  • Robust Polyamide shell with glass fiber-reinforced polypropylene backrest for improved flexibility and strength
  • Plush synthetic leather or durable integral skin foam seat with anti?bacterial properties
  • Class 4 pneumatic cylinder

Options

  • Fixed arms option available
  • Casters: multi-surface dual wheel is standard. Other caster options are also available
  • High-performance twist-lock flat surface chrome footring.Footrings are only available on high-height stools
  • Desk and high height with desk to counter option
  • Durable nylon or polished aluminum base

Specifications

Product Type High Stool
Back Style Standard
With Armrest No
With Casters Yes
Footrest Yes
Color Black, Gray
With Backrest Yes
With Tilt No
With Lumbar Support No
Material Polypropylene
Seating Type Stool with Back
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.