Learn More
Analytical Sales & Services Citus Echelon C18, 4µm, 100mm x 0.075mm (75µm)
Catalog No. 5000807214
Encompass
Citus Echelon C18, 4µm, 100mm x 0.075mm (75µm)
Catalog No. 50-008-07214 Supplier Analytical Sales & Services Supplier No. ET1617
Citus Echelon C18, 4µm, 100mm x 0.075mm (75µm)