Learn More
Analytical Sales & Services Citus Echelon C18, 4µm, 50mm x 0.150mm (150µm)
Catalog No. 5000805298
Encompass
Citus Echelon C18, 4µm, 50mm x 0.150mm (150µm)
Catalog No. 50-008-05298 Supplier Analytical Sales & Services Supplier No. ET1651
Citus Echelon C18, 4µm, 50mm x 0.150mm (150µm)