Learn More
Medchemexpress LLC Ciwujianoside C3 | 114906-74-0 | 1 MG
SDP
Catalog No. 5000418511
Encompass_Preferred
Ciwujianoside C3 is an orally active and brain-penetrated compound isolated from the leaves of Acanthopanax henryi Harms. It exhibits an anti-inflammatory effect and can reinforce object recognition memory.
  • Orally active and brain-penetrated compound
  • Isolated from the leaves of Acanthopanax henryi Harms
  • Exhibits anti-inflammatory effect
  • Can reinforce object recognition memory
Catalog No. 50-004-18511 Supplier Medchemexpress LLC Supplier No. HYN41341MG
Ciwujianoside C3 is an orally active and brain-penetrated compound isolated from the leaves of Acanthopanax henryi Harms. It exhibits an anti-inflammatory effect and can reinforce object recognition memory.
  • Orally active and brain-penetrated compound
  • Isolated from the leaves of Acanthopanax henryi Harms
  • Exhibits anti-inflammatory effect
  • Can reinforce object recognition memory