Learn More
Medchemexpress LLC HY-N4134 5mg Medchemexpress, Ciwujianoside C3 CAS:114906-74-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN41345MG
Encompass_Preferred
Medchemexpress, HY-N4134 5mg Ciwujianoside C3 CAS:114906-74-0 Ciwujianoside C3, an orally active and brain penetrated compound, is isolated the leaves of Acanthopanax henryi Harms. Ciwujianoside C3 has anti-inflammatory effect and can reinforces object recognition memory. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0425