Learn More
Medchemexpress LLC HY-N2175 1mg Medchemexpress, Cixiophiopogon A CAS:288143-27-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21751MG
Encompass_Preferred
Medchemexpress, HY-N2175 1mg Cixiophiopogon A CAS:288143-27-1 Cixiophiopogon A, a steroidal glycoside, obtained from the tuberous roots of Ophiopogon japonicus (Liliaceae). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2872