Learn More
Vitacyte LLC CIzyme RI Lyophilized Collagenase
SDP
Catalog No. NC0110933
Encompass
This item has a minimum qty of 2 per supplier requirements.
CIzyme RI is an aseptically filled, lyophilized mixture of 60% purified class I (C1) and 40% purified class II (C2) collagenase from Clostridium histolyticum pre-mixed with neutral protease from Bacillus polymyxa. Manufactured for superior lot-to-lot enzyme consistency for improved performance consistencyFormulated to contain an optimal amount of Collagen Degrading Activity (CDA) and Neutral Protease Activity (NPA) for rodent islet isolationLow contaminating endotoxin levels (<25 EU/OD)Lot specific Certificate of Analysis provided with complete QC analysis results.
Catalog No. NC0110933 Supplier Vitacyte LLC Supplier No. 0051030
This item has a minimum qty of 2 per supplier requirements.
CIzyme RI is an aseptically filled, lyophilized mixture of 60% purified class I (C1) and 40% purified class II (C2) collagenase from Clostridium histolyticum pre-mixed with neutral protease from Bacillus polymyxa. Manufactured for superior lot-to-lot enzyme consistency for improved performance consistencyFormulated to contain an optimal amount of Collagen Degrading Activity (CDA) and Neutral Protease Activity (NPA) for rodent islet isolationLow contaminating endotoxin levels (<25 EU/OD)Lot specific Certificate of Analysis provided with complete QC analysis results.