Learn More
AdipoGen CJ-21058
SDP
Catalog No. 502562127
Encompass_Preferred
Chemical. CAS 405072-57-3. Formula C23H33NO4. MW 387.5. Isolated from Phoma sp. Antibiotic. SecA inhibitor. Antibacterial and antifungal. Cytotoxic. Methyl analog of equisetin.
Catalog No. 50-256-2127 Supplier AdipoGen Supplier No. BVT0064M001
Add to Cart
Add to Cart
Chemical. CAS 405072-57-3. Formula C23H33NO4. MW 387.5. Isolated from Phoma sp. Antibiotic. SecA inhibitor. Antibacterial and antifungal. Cytotoxic. Methyl analog of equisetin.