Learn More
Abcam CJ-21058, SecA inhibitor, 5MG
Catalog No. 502705972
Encompass_Preferred
MW 387.5 Da, Purity >98%. SecA inhibitor. Shows antiprotozoal effects (IC₅₀ = 7 μM, T. brucei). Inhibits post-translational protein transport into the endoplasmic reticulum. Inhibits ATP-dependant translocation of precursor proteins across a bacterial cell membrane. Shows antibacterial and antifungal effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5972 Supplier Abcam Supplier No. AB1442255MG
MW 387.5 Da, Purity >98%. SecA inhibitor. Shows antiprotozoal effects (IC₅₀ = 7 μM, T. brucei). Inhibits post-translational protein transport into the endoplasmic reticulum. Inhibits ATP-dependant translocation of precursor proteins across a bacterial cell membrane. Shows antibacterial and antifungal effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement