Learn More
Biomiga Inc. CJ236 CHEMICALLY COMPETENT
Catalog No. NC0492648
Encompass_Preferred
The host for preparing ssDNA with a part of Thymine(T) in its DNA replaced with deoxyuracil(dU)
Catalog No. NC0492648 Supplier Biomiga Inc. Supplier No. CC132101
The host for preparing ssDNA with a part of Thymine(T) in its DNA replaced with deoxyuracil(dU)