Learn More
Gold Biotechnology Inc CJ236 Electro E Coli Cells
SDP
Catalog No. 5000562251
Encompass_Preferred
GoldBio CJ236 Electrocompetent E coli cells are ideally suited for use in the Kunkel method of mutagenesis CJ236 cells have part of their Thymine replaced with deoxyuracil in the DNA due to dut - and ung - mutations which prevent the cells from removing uracil from DNA caused by misincorporation or dCTP deamination CJ236 contains the F gene which confers Chloramphenicol resistance
Catalog No. 50-005-62251 Supplier Gold Biotechnology Inc Supplier No. CC2515X50
GoldBio CJ236 Electrocompetent E coli cells are ideally suited for use in the Kunkel method of mutagenesis CJ236 cells have part of their Thymine replaced with deoxyuracil in the DNA due to dut - and ung - mutations which prevent the cells from removing uracil from DNA caused by misincorporation or dCTP deamination CJ236 contains the F gene which confers Chloramphenicol resistance