Learn More
Cayman Chemical CJC-1295 (acetate), Purity: ≥95%, 100 mg
SDP
Catalog No. 502947087
Encompass_Preferred
A synthetic peptide derivative of GHRH; does not contain a DAC moiety, which reduces its half-life; increases the secretion of growth hormone in primary rat anterior pituitary cells when used from pM to µM concentrations; increases the secretion of growth hormone into rat blood at 1 µmol/kg, s.c.
Catalog No. 50-294-7087 Supplier Cayman Chemical Supplier No. 32704100 MG
A synthetic peptide derivative of GHRH; does not contain a DAC moiety, which reduces its half-life; increases the secretion of growth hormone in primary rat anterior pituitary cells when used from pM to µM concentrations; increases the secretion of growth hormone into rat blood at 1 µmol/kg, s.c.