Learn More
Cayman Chemical CJC-1295 (acetate), Purity: ≥95%, 50 mg
SDP
Catalog No. 502949019
Encompass_Preferred
A synthetic peptide derivative of GHRH; does not contain a DAC moiety, which reduces its half-life; increases the secretion of growth hormone in primary rat anterior pituitary cells when used from pM to µM concentrations; increases the secretion of growth hormone into rat blood at 1 µmol/kg, s.c.
Catalog No. 50-294-9019 Supplier Cayman Chemical Supplier No. 3270450 MG
A synthetic peptide derivative of GHRH; does not contain a DAC moiety, which reduces its half-life; increases the secretion of growth hormone in primary rat anterior pituitary cells when used from pM to µM concentrations; increases the secretion of growth hormone into rat blood at 1 µmol/kg, s.c.