Learn More
Cayman Chemical CJC-1295 (acetate), Purity: ≥95%, 5 mg
SDP
Catalog No. 502950987
Encompass_Preferred
A synthetic peptide derivative of GHRH; does not contain a DAC moiety, which reduces its half-life; increases the secretion of growth hormone in primary rat anterior pituitary cells when used from pM to µM concentrations; increases the secretion of growth hormone into rat blood at 1 µmol/kg, s.c.
Catalog No. 50-295-0987 Supplier Cayman Chemical Supplier No. 327045 MG
A synthetic peptide derivative of GHRH; does not contain a DAC moiety, which reduces its half-life; increases the secretion of growth hormone in primary rat anterior pituitary cells when used from pM to µM concentrations; increases the secretion of growth hormone into rat blood at 1 µmol/kg, s.c.