Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cjoc42 | 2171519-89-2 | 99.0% | 415.46 | 100 MG
SDP

Catalog No. 5000348133
Encompass_Preferred

Cjoc42 is a compound capable of binding to gankyrin, an oncoprotein that plays a critical role in cancer development. It inhibits gankyrin activity in a dose-dependent manner, thereby preventing the decrease in p53 protein levels that is often associated with high amounts of gankyrin. This action helps to restore p53-dependent transcription and enhance sensitivity to DNA damage, offering potential as an anti-cancer agent. It targets MDM-2/p53 pathway, associated with apoptosis.

  • Binds to gankyrin and inhibits its activity
  • Restores p53-dependent transcription
  • Enhances sensitivity to DNA damage
  • Recommended storage at 4°C, protected from light
  • Soluble in DMSO

Catalog No. 50-003-48133 Supplier Medchemexpress LLC Supplier No. HY138054100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cjoc42 is a compound capable of binding to gankyrin, an oncoprotein that plays a critical role in cancer development. It inhibits gankyrin activity in a dose-dependent manner, thereby preventing the decrease in p53 protein levels that is often associated with high amounts of gankyrin. This action helps to restore p53-dependent transcription and enhance sensitivity to DNA damage, offering potential as an anti-cancer agent. It targets MDM-2/p53 pathway, associated with apoptosis.

  • Binds to gankyrin and inhibits its activity
  • Restores p53-dependent transcription
  • Enhances sensitivity to DNA damage
  • Recommended storage at 4°C, protected from light
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.