Learn More
Medchemexpress LLC HY-15892 10mg Medchemexpress, CK-636 CAS:442632-72-6 Purity:>98%
SDP
Catalog No. 502081820
Encompass_Preferred
Medchemexpress, HY-15892 10mg CK-636 CAS:442632-72-6 CK-636 is a cell permeable inhibitor of Arp2/3 complex, that could inhibit actin polymerization, with IC50 values of 4 μM, 24 μM and 32 μM for human, fission yeast and bovine, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1820 Supplier Medchemexpress LLC Supplier No. HY1589210MG
Medchemexpress, HY-15892 10mg CK-636 CAS:442632-72-6 CK-636 is a cell permeable inhibitor of Arp2/3 complex, that could inhibit actin polymerization, with IC50 values of 4 μM, 24 μM and 32 μM for human, fission yeast and bovine, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.