Learn More
TARGETMOL CHEMICALS INC CK-636 50MG
SDP
Catalog No. 502577293
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CK-636 (CK-0944636) an Arp2/3 complex inhibitor can inhibit actin polymerization induced by the human (IC50 4 uM) fission yeast (IC50 24 uM) and bovine (IC50 32 uM) Arp2/3 complex respectively. purity: 98%
Catalog No. 50-257-7293 Supplier TARGETMOL CHEMICALS INC Supplier No. T182050MG
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CK-636 (CK-0944636) an Arp2/3 complex inhibitor can inhibit actin polymerization induced by the human (IC50 4 uM) fission yeast (IC50 24 uM) and bovine (IC50 32 uM) Arp2/3 complex respectively. purity: 98%