Learn More
Enzo Life Sciences CK-666 (2mg). CAS: 442633-00-3
SDP
Catalog No. 502008629
Encompass_Preferred
Cell permeable inhibitor human (IC50=4µM), fission yeast (IC50=5µM), S. cerevisiae (IC50=12µM) and bovine (IC50=17µM) Arp2/3 complex between Arp2 and Arp3. Inhibits Listeria motility (IC50=7µM). Alternative name: 2-Fluoro-N-[2-(2-methyl-1H-indol-3-yl)ethyl]-benzamide. Purity: ≥95%. Solubility: Soluble in DMSO (a minimum concentration of 5 mg/ml is recommended). Long Term Storage: -20°C.
Catalog No. 50-200-8629 Supplier Enzo Life Sciences Supplier No. ALX270506M002
Cell permeable inhibitor human (IC50=4µM), fission yeast (IC50=5µM), S. cerevisiae (IC50=12µM) and bovine (IC50=17µM) Arp2/3 complex between Arp2 and Arp3. Inhibits Listeria motility (IC50=7µM). Alternative name: 2-Fluoro-N-[2-(2-methyl-1H-indol-3-yl)ethyl]-benzamide. Purity: ≥95%. Solubility: Soluble in DMSO (a minimum concentration of 5 mg/ml is recommended). Long Term Storage: -20°C.