Learn More
Selleck Chemical LLC CK-666-5mg
SDP
Catalog No. 502389772
Encompass_Preferred
CK-666 is an inhibitor of the actin-related protein 2/3 (Arp2/3) complex with IC50 of 17 μM and 5 μM for BtArp2/3 complex and SpArp2/3 complex, respectively.
Catalog No. 50-238-9772 Supplier Selleck Chemical LLC Supplier No. S76905MG
CK-666 is an inhibitor of the actin-related protein 2/3 (Arp2/3) complex with IC50 of 17 μM and 5 μM for BtArp2/3 complex and SpArp2/3 complex, respectively.