Learn More
Medchemexpress LLC HY-16926 5mg Medchemexpress, CK-666 CAS:442633-00-3 Purity:>98%
SDP
Catalog No. 501932785
Encompass_Preferred
Medchemexpress, HY-16926 5mg CK-666 CAS:442633-00-3 CK-666 is a cell-permeable inhibitor of actin-related protein Arp2/3 complex , and binds to Arp2/3 complex, stabilizes the inactive state of the complex, blocking movement of the Arp2 and Arp3 subunits into the activated filament-like (short pitch) conformation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2785 Supplier Medchemexpress LLC Supplier No. HY169265MG
Medchemexpress, HY-16926 5mg CK-666 CAS:442633-00-3 CK-666 is a cell-permeable inhibitor of actin-related protein Arp2/3 complex , and binds to Arp2/3 complex, stabilizes the inactive state of the complex, blocking movement of the Arp2 and Arp3 subunits into the activated filament-like (short pitch) conformation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.