Learn More
Abcam CK 666, Actin polymerization inhibitor, 10MG
Catalog No. 502583365
Encompass_Preferred
MW 296.3 Da, Purity >99%. Potent inhibitor of actin polymerization. Inhibits the Arp2/3 (IC₅₀ = 17 μM), SpArp2/3 (IC₅₀ = 5 μM) and HsArp2/3 complexes (IC₅₀= 4 μM) by preventing a shift into an active conformation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3365 Supplier Abcam Supplier No. AB14123110MG
MW 296.3 Da, Purity >99%. Potent inhibitor of actin polymerization. Inhibits the Arp2/3 (IC₅₀ = 17 μM), SpArp2/3 (IC₅₀ = 5 μM) and HsArp2/3 complexes (IC₅₀= 4 μM) by preventing a shift into an active conformation.
The product is subject to the following: Abcam Restricted Use Statement