Learn More
Genscript Corporation CK-MB 8D8HC
Catalog No. 5000058412
Encompass
Creatine kinase (CK) is a dimeric enzyme including three isoforms of creatine kinase BB MM and MB Level of serum CK-MB will be increased within 4-6 hours after a myocardial infarction (MI) CK-MB is considered as a useful marker for the diagnosis of myocardial infarction
Catalog No. 50-000-58412 Supplier Genscript Corporation Supplier No. V0130410
Creatine kinase (CK) is a dimeric enzyme including three isoforms of creatine kinase BB MM and MB Level of serum CK-MB will be increased within 4-6 hours after a myocardial infarction (MI) CK-MB is considered as a useful marker for the diagnosis of myocardial infarction