Learn More
Sino Biological Recombinant Human CSNK1G1 Protein (GST Tag), Active
Catalog No. 502575473
Encompass_Preferred
Recombinant human CK1 gamma 1 (21-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5473 Supplier Sino Biological Supplier No. C6811G50
Recombinant human CK1 gamma 1 (21-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.