Learn More
Sino Biological Recombinant Human CSNK1E/CK1 epsilon Protein (Full Length, GST Tag), Active
Catalog No. 502569497
Encompass_Preferred
Recombinant full length human CK1 epsilon was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-256-9497 Supplier Sino Biological Supplier No. C6610G10
Recombinant full length human CK1 epsilon was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.