Learn More
Sino Biological Recombinant Human Casein Kinase 1 alpha Protein (Full Length, GST Tag), Active
Catalog No. 502575488
Encompass_Preferred
Recombinant full length human CK1 alpha 1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5488 Supplier Sino Biological Supplier No. C6410G500
Recombinant full length human CK1 alpha 1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.