Learn More
Medchemexpress LLC HY-111820 10mg Medchemexpress, CK1-IN-1 CAS:1784751-20-7 Purity:>98%
SDP
Catalog No. 501968974
Encompass_Preferred
Medchemexpress, HY-111820 10mg CK1-IN-1 CAS:1784751-20-7 CK1-IN-1 is a casein kinase 1 (CK1) inhibitor extracted from patent WO2015119579A1, compound 1c, has IC50s of 15 nM, 16 nM, 73 nM for CK1δ, and CK1ε, p38σ MAPK, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8974 Supplier Medchemexpress LLC Supplier No. HY11182010MG
Medchemexpress, HY-111820 10mg CK1-IN-1 CAS:1784751-20-7 CK1-IN-1 is a casein kinase 1 (CK1) inhibitor extracted from patent WO2015119579A1, compound 1c, has IC50s of 15 nM, 16 nM, 73 nM for CK1δ, and CK1ε, p38σ MAPK, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.