Learn More
Carnabio Usa Inc CK1E(CSNK1E)/5UG/CATALYTIC
Catalog No. 507540379
Encompass
CK1(CSNK1E), Catalytic domain, Wild type, Amino Acid 1-348, NP_001885.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0379 Supplier Carnabio Usa Inc Supplier No. 031045
CK1(CSNK1E), Catalytic domain, Wild type, Amino Acid 1-348, NP_001885.1, Expressed in Insect (sf21); 5microg