Learn More
Sino Biological Recombinant Human Casein Kinase 1 gamma 2 Protein (Full Length, GST Tag), Active
Catalog No. 502575477
Encompass_Preferred
Recombinant full length human CK1 gamma 2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5477 Supplier Sino Biological Supplier No. C6810BG50
Recombinant full length human CK1 gamma 2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.