Learn More
TARGETMOL CHEMICALS INC CK2/ERK8-IN-1 5MG
SDP
Catalog No. 502581652
Encompass_Preferred
Also available in 10 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CK2/ERK8-IN-1 (TMCB) is a dual inhibitor of casein kinase 2 (CK2) (Ki 0.25 µM) and ERK8 (IC50s 0.50 uM) with pro-apoptotic efficacy. CK2/ERK8-IN-1 also binds to PIM1 DYRK1A and HIPK2 (Kis 8.65 µM 11.9 µM and 15.25 µM). purity: 98%
Catalog No. 50-258-1652 Supplier TARGETMOL CHEMICALS INC Supplier No. T108275MG
Also available in 10 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CK2/ERK8-IN-1 (TMCB) is a dual inhibitor of casein kinase 2 (CK2) (Ki 0.25 µM) and ERK8 (IC50s 0.50 uM) with pro-apoptotic efficacy. CK2/ERK8-IN-1 also binds to PIM1 DYRK1A and HIPK2 (Kis 8.65 µM 11.9 µM and 15.25 µM). purity: 98%