Learn More
AdipoGen CK2 Inhibitor 10 (5 mg)
SDP
Supplier:  AdipoGen AGCR13626M005
Encompass_Preferred
Chemical. CAS: 1361229-76-6. Formula: C18H14N2O4S . 0.5H2O. MW: 354.4 . 9.0. Potent and ATP-competitive inhibitor of casein kinase 2 (IC50 CK2alpha=32nM and IC50 CK2alpha'=46nM). Potently also inhibits DYRK1B and FLT3. Anticancer compound. Exhibits potent cytotoxicity towards lung cancer cells A549, colorectal cancer cells HCT-116 and breast cancer cells MCF-7. Promoted cAMP-induced thermogenesis in white adipocytes. CK2 inhibition ameliorates diet-induced obesity and insulin resistance in mice in vivo by promoting UCP1-dependent thermogenesis.
Catalog No. 50-205-3996