Learn More
Medchemexpress LLC CK2 inhibitor 2 | 2641079-92-5 | 98.5% | C21H17ClN4O2 | 10MG
SDP
Catalog No. 5000260754
Encompass_Preferred
CK2 inhibitor 2 is a potent, selective, orally active small-molecule inhibitor of casein kinase 2 (CK2) with sub-nanomolar enzymatic potency and documented cellular activity. Supplied as a solid for research use, it includes solubility data for DMSO and recommended storage conditions for powder and solutions.
  • Potent CK2 inhibition (IC50 0.66 nM) for biochemical assays.
  • Selectable kinase profile with measurable activity against Clk2.
  • Cellular activity in multiple cancer cell lines at low micromolar concentrations.
  • High purity suitable for biochemical and cell-based studies (98.5%).
  • Documented solubility in DMSO and example in vivo formulation options.
  • Defined storage recommendations for powder and solvent preparations.
Catalog No. 50-002-60754 Supplier Medchemexpress LLC Supplier No. HY13217510MG
CK2 inhibitor 2 is a potent, selective, orally active small-molecule inhibitor of casein kinase 2 (CK2) with sub-nanomolar enzymatic potency and documented cellular activity. Supplied as a solid for research use, it includes solubility data for DMSO and recommended storage conditions for powder and solutions.
  • Potent CK2 inhibition (IC50 0.66 nM) for biochemical assays.
  • Selectable kinase profile with measurable activity against Clk2.
  • Cellular activity in multiple cancer cell lines at low micromolar concentrations.
  • High purity suitable for biochemical and cell-based studies (98.5%).
  • Documented solubility in DMSO and example in vivo formulation options.
  • Defined storage recommendations for powder and solvent preparations.