Learn More
Newcomer Supply CK5/14 + p63 + P504S, Multi-Tissue Control Slides, 98/set
SDP
Catalog No. NC0749117
Encompass
Newcomer Supply CK5/14 + p63 + P504S, Multi-Tissue Control Slides provide a useful combination of markers helpful in diagnosing prostatic intraepithelial neoplasia (PIN). CK5/14 and P504S contain sections of positive staining prostate and p63 contains positive staining prostate and negative staining myometrium.
Catalog No. NC0749117 Supplier Newcomer Supply Supplier No. 3650B
Newcomer Supply CK5/14 + p63 + P504S, Multi-Tissue Control Slides provide a useful combination of markers helpful in diagnosing prostatic intraepithelial neoplasia (PIN). CK5/14 and P504S contain sections of positive staining prostate and p63 contains positive staining prostate and negative staining myometrium.