Learn More
Newcomer Supply CK5/14 + p63 + P504S (PIN-4), Multi-Tissue Control Slides, 10/set
SDP
Supplier:  Newcomer Supply 3650A
Encompass
Newcomer Supply CK5/14 + p63 + P504S, Multi-Tissue Control Slides provide a useful combination of markers helpful in diagnosing prostatic intraepithelial neoplasia (PIN). P504S and CK5/14 contain sections of positive staining prostate and p63 contains sections of positive staining prostate and negative staining myometrium.
Catalog No. NC2101949