Learn More
Enzo Life Sciences Cytokeratin 8, 18 & 19 monoclonal antibody (2A4) (100µl)
SDP
Supplier:  Enzo Life Sciences BMLCA11410100
Encompass_Preferred
Clone: 2A4. Isotype: IgG1. Immunogen: Cytokeratins isolated from cultured porcine kidney epithelial cells (LLC-PK). Species reactivity: Human, Mouse, Rat Porcine. Specificity: Recognizes cytokeratin proteins 8, 18 and 19. Applications: ICC, WB. Purity: Protein A affinity chromatography. Formulation: Liquid. In PBS containing 1% BSA and 0.09% sodium azide.
Catalog No. 50-201-0359