Learn More
Abcam CK869, Actin-related protein 2/3 (Arp2/3) comple x inhibitor, 10MG
Catalog No. 502672829
Encompass_Preferred
MW 394.3 Da, Purity >98%. Inhibitor of human and bovine actin-related protein 2/3 (Arp2/3) complex. Inhibits actin polymerization with bovine Arp2/3 complex (IC₅₀ = 11 μM) and inhibits the formation of actin filament comet tails by Listeriamonocytogenes in infected SKOV3 cells (IC₅₀ = 7 μM) . Does not inhibit budding or fission yeast Arp2/3 complexes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2829 Supplier Abcam Supplier No. AB21810910MG
MW 394.3 Da, Purity >98%. Inhibitor of human and bovine actin-related protein 2/3 (Arp2/3) complex. Inhibits actin polymerization with bovine Arp2/3 complex (IC₅₀ = 11 μM) and inhibits the formation of actin filament comet tails by Listeriamonocytogenes in infected SKOV3 cells (IC₅₀ = 7 μM) . Does not inhibit budding or fission yeast Arp2/3 complexes.
The product is subject to the following: Abcam Restricted Use Statement