Learn More
Aviva Systems Biology Corp CKAP4 Antibody - middle region (ARP46790_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46790_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CKAP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CKAP4. Alternate names: CLIMP-63, ERGIC-63, MGC99554, p63ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59400